Learn More
Medchemexpress LLC 6-Bromo-2-pyridinecarboxaldehyde | 34160-40-2 | 99.7% | C6H4BrNO | 10 G
SDP
Supplier:  Medchemexpress LLC HY6019410G
Encompass_Preferred
6-Bromo-2-pyridinecarboxaldehyde is a biochemical reagent suitable for various life science research applications, acting as a biological material or organic compound. It is specifically utilized in the study of rhodium-catalyzed reductive aldol coupling with divinyl ketones, which results in syn-hydroxyenones. Additionally, this compound functions as a foundational building block for Tris[(pyridyl)methyl]amine ligands.
  • Functions as a biological material for life science research
  • Acts as an organic compound for life science research
  • Utilized in rhodium-catalyzed reductive aldol coupling with divinyl ketones
  • Leads to the formation of syn-hydroxyenones
  • Serves as a building block for Tris[(pyridyl)methyl]amine ligands
Catalog No. 50-004-34199