Learn More
Medchemexpress LLC 6-Bromo-4-chloroquinoline | 65340-70-7 | 100.0% | C9H5BrClN | 100 G

Supplier: Medchemexpress LLC HY75144100G
6-Bromo-4-chloroquinoline is a drug intermediate used for the synthesis of various active compounds. It is a solid with a white to light yellow color. It has a molecular weight of 242.50. This product is for research use only.
- Used for the synthesis of various active compounds.
- Solid form.
- White to light yellow color.
- Molecular weight of 242.50.
- Store at room temperature for 3 years.
- Store in solvent at -80°C for 2 years or -20°C for 1 year.
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.