Learn More
Medchemexpress LLC 6-Bromo-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole | 1231930-33-8 | MFCD16660227 | C11H12BrFN2 | 500 G
SDP
Supplier:  Medchemexpress LLC HY78066500G
Encompass_Preferred
Abemaciclib Impurity 1 (6-Bromo-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole) is a drug intermediate used for the synthesis of various active compounds. It is intended for use as a laboratory chemical and in the manufacture of substances. This product is for research use only and has not been fully validated for medical applications.
  • Functions as a drug intermediate
  • Appears as a solid
  • Molecular weight of 271.13
  • Stable under recommended storage conditions
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
Catalog No. 50-004-34855