Learn More
Medchemexpress LLC 6-Bromo-L-tryptophan | 52448-17-6 | 99.7% | 283.13 | 1 G
SDP
Supplier:  Medchemexpress LLC HY208991G
Encompass_Preferred
6-Bromo-L-tryptophan (6BrW) is a compound that can be incorporated into recombinant proteins by a specific system in bacterial cells, causing a blue shift in fluorescence emission and absorption maxima. It is for research use only.
  • Can be incorporated into recombinant proteins.
  • Causes a blue shift in fluorescence emission and absorption maxima.
  • For research use only.
Catalog No. 50-004-00700