Learn More
TARGETMOL CHEMICALS INC 6-Bromoindole-3-acetonitrile 2
SDP
Supplier:  TARGETMOL CHEMICALS INC TN719825MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 6-Bromoindole-3-acetonitrile (2-(6-bromo-1H-indol-3-yl)acetonitrile) is a marine derived natural products found in Pseudosuberites hyalinus.
Catalog No. 50-287-9351