Learn More
Medchemexpress LLC 6-Bromoindole | 52415-29-9 | 99.9% | C8H6BrN | 50 G

Supplier: Medchemexpress LLC HY5923350G
6-Bromoindole is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Used as a biological material
- Used as an organic compound
- Suitable for life science related research
- Shipping at room temperature
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.