Learn More
Cayman Chemical 6-CarboxyfluoresceIn 100mg
SDP
Supplier:  Cayman Chemical 21233100 MG
Encompass_Preferred
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively
Catalog No. 50-298-5428