Learn More
Medchemexpress LLC 6-CEPN 5mg | 1054549-73-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1145695MG
Encompass_Preferred
6-CEPN is a small-molecule RAS inhibitor that binds Icmt sites, showing anticancer activity by blocking cells in the G1 phase and inducing autophagy and necrosis. Supplied for research use in small-mass packs.
  • Inhibits RAS activation by targeting Icmt binding sites.
  • Induces autophagy and necrosis in cancer cells.
  • Useful for studies of ras signaling, autophagy, and cell-cycle regulation.
  • Provided in small-mass packs suitable for in vitro assays.
  • Soluble in DMSO at 100 mg/mL; formulation guidance provided for in vivo dosing vehicles.
Catalog No. 50-002-29548