Learn More
Medchemexpress LLC 6-chloro-2,3,4,5-tetrahydro-1-phenyl-1H-3-benzazepine-7,8-diol, monohydrobromide | 67287-39-2 | >=98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1223650MG
Encompass_Preferred
SKF 81297 hydrobromide is a selective dopamine D1 receptor agonist supplied as the hydrobromide salt for research use. It is used in neuropharmacology to probe D1-like receptor function in vitro and in vivo and is available in multiple research-scale pack sizes.
  • Selective dopamine D1 receptor agonist.
  • Supplied as the hydrobromide salt for research use.
  • High purity suitable for research (≥98%).
  • Molecular weight 370.67 g/mol; formula C16H17BrClNO2.
  • Available in multiple pack sizes including milligram and solution formats.
Catalog No. 50-002-31282