Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Chloro-2-fluoro-9H-purine | 1651-29-2 | 172.55 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW00292410G

Encompass_Preferred

6-Chloro-2-fluoro-9H-purine is a biochemical reagent that can be used as a biological material or organic compound for various life science related research applications.

  • Utilized as a biochemical reagent
  • Can be used as a biological material or organic compound
  • Solid appearance, light yellow to yellow in color
  • Molecular formula: C5H2ClFN4
  • Purity: 99.2%
  • Molecular weight: 172.55
  • CAS number: 1651-29-2
  • Melting/freezing point: 162-163°C
  • Boiling point/range: 553.2°C at 760 mmHg
  • Relative density: 1.98 g/cm³
  • Recommended storage at -20°C, sealed, away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-48690


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.