Learn More
Medchemexpress LLC 6-chloro-3-[(1E)-3-[[(2-methoxyphenyl)methyl]amino]-3-oxo-1-propen-1-yl]-1H-indole-2-carboxylic | 2349374-37-2 | 98.8% | 412.87 g·mol⁻¹ | C22H21ClN2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1389895MG
Encompass_Preferred
15-LOX-1 inhibitor 1 is a small-molecule research reagent that selectively inhibits 15-lipoxygenase-1 (15-LOX-1) with an IC50 of 0.19 μM. It has been reported to protect macrophages from lipopolysaccharide-induced cytotoxicity and to inhibit nitric oxide formation and lipid peroxidation. The compound is supplied as a solid and as a DMSO solution in multiple sizes for laboratory use.
  • Inhibits 15-lipoxygenase-1 (IC50 = 0.19 μM).
  • Protects macrophages from LPS-induced cytotoxicity.
  • Reduces nitric oxide formation and lipid peroxidation.
  • High purity suitable for research (98.8%).
  • Available as solid and as a DMSO solution in multiple sizes.
Catalog No. 50-002-23318