Learn More
Medchemexpress LLC 6-chloro-4-(cyclopentylamino)pyrazolo[3,4-d]pyrimidinyl glycosyl phosphonic acid | 2216764-29-1 | 99.2% | C16H23ClN5O7P | 10MG
SDP
Supplier:  Medchemexpress LLC HY13196710MG
Encompass_Preferred
CD73-IN-4 is a potent, selective methylenephosphonic acid inhibitor of CD73 used in preclinical research on cancer immunology. It is supplied as a high-purity solid and includes solubility and storage guidance for in vitro and in vivo studies.
  • Potent CD73 inhibition (IC50 = 2.6 nM for human CD73).
  • High purity suitable for research (99.2%).
  • Defined chemical formula and molecular weight for assay planning (C16H23ClN5O7P; 463.81 g·mol⁻¹).
  • Good solubility in DMSO (125 mg/mL) for in vitro use; in vivo formulation protocols available.
  • Stable when stored as powder at -20°C for long-term use.
Catalog No. 50-002-60956