Learn More
Medchemexpress LLC 6-chloro-9H-purine | 87-42-3 | MFCD00075825 | 99.5% | 154.56 | C5H3ClN4 | 50g
SDP
Supplier:  Medchemexpress LLC HYY024750G
Encompass_Preferred
6-Chloropurine is a building block in chemical synthesis Intermediate in the preparation of 9-alkylpurines and 6-rnercaptopurine Antitumor activities[1]
Catalog No. 50-002-66539