Learn More
AdipoGen 6-Chloropurine (5 g)
SDP
Supplier:  AdipoGen CDXC0684G005
Encompass_Preferred
Chemical. CAS: 87-42-3. Formula: C5H3ClN4. MW: 154.56. 6-Chloropurine is a building block in chemical synthesis and is used as an intermediate in the preparation of 9-alkylpurines via alkylation with various substituted alkyl halides in DMSO. 6-Chloropurine based compounds have been shown to display antiviral and antitumor properties.
Catalog No. 50-205-4344