Learn More
Medchemexpress LLC 6-Demethoxytangeretin | 6601-66-7 | 5 MG

Supplier: Medchemexpress LLC HYN41265MG
6-Demethoxytangeretin is a flavonoid compound isolated from *Citrus reticulata*. It exhibits anti-inflammatory and anti-allergic properties, inhibiting the production of IL-6 and the expression of related genes in human mast cells through the ALK and MAPK pathways. Additionally, it can promote CRE-mediated transcription in hippocampal neurons.
- Anti-inflammatory and anti-allergic activities
- Neuronal activity
- Naturally isolated from *Citrus reticulata*
- Purity: 99.37%
- Appearance: solid, white to off-white color
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.