Learn More
Cayman Chemical 6-deoxy-L-Talose 10mg
SDP
Supplier:  Cayman Chemical 2091110 MG
Encompass_Preferred
A microbial monosaccharide; has been found in the cell wall of bacteria such as S. bovis and in the O-specific polysaccharide of LPS in species including E. coli and P. maltophilia; a component of the surface glycopeptidolipids of M. avium
Catalog No. 50-298-3365