Learn More
Medchemexpress LLC HY-108357 5mg Medchemexpress, 6-Diazo-5-oxo-L-nor-Leucine CAS:157-03-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1083575MG
Encompass_Preferred
Medchemexpress, HY-108357 5mg 6-Diazo-5-oxo-L-nor-Leucine CAS:157-03-9 L-6-Diazo-5-oxonorleucine (L-6-Diazo-5-oxonorleucine) is a glutaminases antagonist with a Ki of 6 μM. L-6-Diazo-5-oxonorleucine exhibits analgesic, antibacterial, antiviral and anticancer properties. L-6-Diazo-5-oxonorleucine displays genetic toxicity in vitro. L-6-Diazo-5-oxonorleucine decreases the self-renewal potential and metastatic ability of tumor cell[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8537