Learn More
TARGETMOL CHEMICALS INC 6-Diazo-5-oxo-L-nor-Leucine
SDP
Supplier:  TARGETMOL CHEMICALS INC T83735MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 6-Diazo-5-oxo-L-nor-Leucine (L-6-Diazo-5-oxonorleucine) is a glutaminase antagonist (Ki 6 uM) and an antineoplastic antibiotic produced by an unidentified Streptomyces species from Peruvian soil. Purity 98.93%
Catalog No. 50-287-6849