Learn More
Apexbio Technology LLC 6-FAM 3301-79-9 500mg
SDP
Supplier:  Apexbio Technology LLC A4666500
Encompass_Preferred
6-FAM (3301-79-9) is a fluorescent dye containing a carboxyl functional group capable of forming covalent linkages with primary amines 6-FAM is utilized as a labeling reagent for biomolecules such as nucleic acids and proteins Based on these properties 6-FAM holds research potential in nucleic acid sequencing fluorescence-based assays for enzyme activity assessment and investigations of cellular signaling pathways in molecular biology and biomedical research
Catalog No. 50-000-94947