Learn More
STA PHARMACEUTICAL US LLC 6-FAM amidite | 1 g | CAS 204697-37-0 | MDL MFCD01940925
Supplier: STA PHARMACEUTICAL US LLC STAAM101051GM
6-FAM amidite is a Amidite reagent (Subcategory: Dye Amidite) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 204697-37-0
- MDL: MFCD01940925
- InChIKey: MGPYJVWEJNTXLC-UHFFFAOYSA-N
- Molecular Weight: 843.954761998
- Molecular Formula: C46H58N3O10P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29322090
- Country Of Origin: China
- IUPAC: 6-((6-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)hexyl)carbamoyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl bis(2,2-dimethylpropanoate)
- SMILES: CC(C)N(C(C)C)P(OCCCCCCNC(C1=CC2=C(C(OC23C4=C(OC5=C3C=CC(OC(C(C)(C)C)=O)=C5)C=C(OC(C(C)(C)C)=O)C=C4)=O)C=C1)=O)OCCC#N
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.