Learn More
Medchemexpress LLC 6'''-Feruloylspinosin | 77690-92-7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN21605MG
Encompass_Preferred
6'''-Feruloylspinosin is a flavonoid isolated from seeds of Ziziphus jujuba. It can cross the blood-brain barrier and enhance the expression of GABAAα1, GABAAα5, and GABABR1 mRNA in rat hippocampal neurons.
  • Isolated from seeds of Ziziphus jujuba
  • Crosses the blood-brain barrier
  • Enhances expression of GABAAα1, GABAAα5, and GABABR1 mRNA
  • Classified as a flavonoid, flavone, phenol, and polyphenol
  • Appearance: Solid
  • Color: Off-white to yellow
Catalog No. 50-004-20247