Learn More
Medchemexpress LLC 6-fluoro-3-[(1E)-2-(3-pyridinyl)ethenyl]-1H-indole | 163239-22-3 | 99.5% | 238.26 g/mol | C15H11FN2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1086815MG
Encompass_Preferred
680C91 is an orally active, selective small-molecule inhibitor of tryptophan 2,3-dioxygenase (TDO) used as a research reagent to probe tryptophan catabolism and its role in cancer immunotherapy and neurodegenerative disease. It is reported with a Ki of 51 nM, molecular formula C15H11FN2, and is supplied as a powder for laboratory use.
  • Selective TDO inhibition (Ki ~51 nM)
  • Orally active small molecule suitable for in vivo and in vitro studies
  • Molecular weight 238.26 g/mol, formula C15H11FN2
  • High reported purity on manufacturer product page
  • Supplied as a powder with recommended cold storage
  • Applicable to cancer immunotherapy and neurodegenerative disease research
Catalog No. 50-002-64296