Learn More
Cayman Chemical 6-fluoro-DL-Tryptophn 50mg
SDP
Supplier:  Cayman Chemical 1764350 MG
Encompass_Preferred
A serotonin synthesis inhibitor that is used for tracing pools of neuronal serotonin, as a competitive inhibitor of tryptophan binding to albumin, and as a substrate for the serotonin neural transporter
Catalog No. 50-296-6097