Learn More
TARGETMOL CHEMICALS INC 6-GNTI DIHYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T399885MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. 6'-GNTI dihydrochloride is a (kappa)-opioid receptor (KOR) agonist that favors activation of G-protein-mediated signaling over recruitment of (beta)-aspirin 2. 6'-GNTI dihydrochloride only activates the Akt pathway in striatal neurons. purity: 98%
Catalog No. 50-258-0024