Learn More
Medchemexpress LLC 6-Hepten-1-ol 25g
SDP
Supplier:  Medchemexpress LLC 25GHYW127475
Encompass_Preferred
6-Hepten-1-ol has a floral and green odor and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of perfumes air fresheners and cleaning agents
Catalog No. 50-002-06881