Learn More
Medchemexpress LLC 6-hexadecenoic acid, sodium salt (1:1), (6Z)- | 217477-25-3 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY130187A10MM/1ML
Encompass_Preferred
Sapienic acid sodium is a fatty acid commonly found on the skin and in mucosa, possessing variable antimicrobial activities against both Gram-positive and Gram-negative bacteria. It is active against Streptococcus sanguinis, Streptococcus mitis, and Fusobacterium nucleatum.
  • Found naturally on skin and mucosa.
  • Exhibits antimicrobial activity against various bacteria.
  • Active against specific Gram-positive and Gram-negative bacteria.
  • Soluble in ethanol for in vitro applications.
Catalog No. 50-003-39046