Learn More
Cayman Chemical 6-hydroxy Etodolac 10mg
SDP
Supplier:  Cayman Chemical 3421110 MG
Encompass_Preferred
A metabolite of etodolac; induces false-positive reactions in diazo diagnostic tests for bilirubin in urine
Catalog No. 50-294-7318