Learn More
Cayman Chemical 6-hydroxy Etodolac 5mg
SDP
Supplier:  Cayman Chemical 342115 MG
Encompass_Preferred
A metabolite of etodolac; induces false-positive reactions in diazo diagnostic tests for bilirubin in urine
Catalog No. 50-295-0701