Learn More
Medchemexpress LLC 6-hydroxy-L-DOPA | 27244-64-0 | >95.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1219395MG
Encompass_Preferred
6-Hydroxy-L-DOPA is an analytical standard and biochemical assay reagent used for research into L-DOPA metabolism and analytical method development. It is a hydroxylated derivative of L-DOPA with molecular formula C9H11NO5 and molecular weight 213.19 g/mol.
  • Analytical standard for chromatographic and mass spectrometric assays.
  • Suitable as a reference material for impurity and metabolite studies.
  • Available in small pack sizes suitable for analytical use.
  • Purity typically >95% as documented by a certificate of analysis.
  • Supplied as a solid; store under conditions recommended by the supplier.
Catalog No. 50-002-30541