Learn More
Medchemexpress LLC 6α-hydroxy paclitaxel-d5 | 1315376-90-9 | MFCD34574651 | 99.3% | 874.94 | C47H46D5NO15 | 1MG
SDP
Supplier:  Medchemexpress LLC HY125348S1MG
Encompass_Preferred
6α-Hydroxy Paclitaxel-d5 is the deuterium-labeled form of a primary human metabolite of paclitaxel, supplied for research use as an isotope-labeled internal standard for quantitative analysis and pharmacokinetic/metabolism studies. Typical packaging is 1 mg and the product is provided with batch COA and analytical data for traceability.
Catalog No. 50-002-37258