Learn More
Medchemexpress LLC HY-78961 50mg Medchemexpress, 6-Maleimidohexanoic acid N-hydroxysuccinimide ester CAS:55750-63-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY7896150MG
Encompass_Preferred
Medchemexpress, HY-78961 50mg 6-Maleimidohexanoic acid N-hydroxysuccinimide ester CAS:55750-63-5 6-Maleimidohexanoic acid N-hydroxysuccinimide ester (EMCS) is a heterobifunctional cross-linking reagent. EMCS is used as a unique and useful reagent for preparation of hapten conjugate and enzyme immunoconjugates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2292