Learn More
Medchemexpress LLC 6-maleimidohexanoic acid N-hydroxysuccinimide ester | 55750-63-5 | 100.0% | 308.29 g·mol⁻1 | C14H16N2O6 | 5 G
SDP
Supplier:  Medchemexpress LLC HY789615G
Encompass_Preferred
6-maleimidohexanoic acid N-hydroxysuccinimide ester (EMCS) is a heterobifunctional cross-linking reagent containing an N-hydroxysuccinimide (NHS) ester reactive toward primary amines and a maleimide group reactive toward thiols. It is used to prepare hapten conjugates and enzyme-immunoconjugates in bioconjugation workflows.
  • Heterobifunctional NHS-ester and maleimide for selective conjugation.
  • High purity suitable for sensitive bioconjugation (reported 99.98%).
  • Effective for linking amine-containing molecules to sulfhydryl-containing molecules.
  • Available in multiple pack sizes, including 5 G.
Catalog No. 50-002-02367