Learn More
Medchemexpress LLC 6-maleimidohexanoic acid N-hydroxysuccinimide ester | 55750-63-5 | 100.0% | 308.29 g/mol | C14H16N2O6 | 25 G

Supplier: Medchemexpress LLC HY7896125G
6-Maleimidohexanoic acid N-hydroxysuccinimide ester is a heterobifunctional cross-linking reagent used to conjugate sulfhydryl groups to amines or carboxyl-functionalized molecules. It is non-cleavable and supplied as a high-purity solid with recommended sealed storage at low temperature.
- Heterobifunctional cross-linker for sulfhydryl-to-amine conjugation.
- High purity suitable for sensitive bioconjugation applications.
- Molecular weight 308.29 g/mol and formula C14H16N2O6.
- CAS number 55750-63-5 for unambiguous identification.
- Recommended storage: -20°C, sealed, away from moisture and light; in solvent: -80°C for long-term.
- Available in multiple pack sizes including 25 G for scale-up.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.