Learn More
Medchemexpress LLC 6-maleimidohexanoic acid N-hydroxysuccinimide ester | 55750-63-5 | >98.0% | 308.29 g/mol | C14H16N2O6 | 10 G

Supplier: Medchemexpress LLC HY7896110G
6-Maleimidohexanoic acid N-hydroxysuccinimide ester (EMCS) is a heterobifunctional cross-linking reagent that couples amine-containing molecules to sulfhydryl groups via an NHS ester and a maleimide. It is used for hapten-carrier conjugation, enzyme immunoconjugates, and antibody-drug conjugate linker chemistry. For research use only.
- Reactive groups: maleimide for sulfhydryl coupling, NHS ester for amine coupling.
- High purity suitable for bioconjugation (≥98.0% by HPLC).
- Molecular weight 308.29 g/mol; formula C14H16N2O6.
- Available in laboratory pack sizes including 10 g.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.