Learn More
Medchemexpress LLC 6-maleimidohexanoic acid N-hydroxysuccinimide ester | 55750-63-5 | MFCD00043043 | 100.0% | 308.29 | C14H16N2O6 | 100 G
SDP
Supplier:  Medchemexpress LLC HY78961100G
Encompass_Preferred
6-maleimidohexanoic acid N-hydroxysuccinimide ester (EMCS) is a heterobifunctional cross-linking reagent that contains a maleimide group for selective thiol coupling and an N-hydroxysuccinimide (NHS) ester for amine coupling. It is used to prepare hapten conjugates and enzyme immunoconjugates, enabling formation of stable thioether and amide linkages.
  • Heterobifunctional reagent for thiol and amine coupling.
  • Facilitates preparation of hapten and enzyme immunoconjugates.
  • Enables formation of stable thioether and amide linkages.
  • High purity suitable for bioconjugation applications.
  • Available in multiple pack sizes, including 100 g.
Catalog No. 50-002-02743