Learn More
Medchemexpress LLC 6-mercaptopurine-13C,15N2 hydrochloride | 207729-57-5 | 99.9% | 191.62 | C_4_^13^CH_5_ClN_2_^15^N_2_S | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW6542581MG
Encompass_Preferred
6-Mercaptopurine-13C,15N2 hydrochloride is a stable isotope-labeled form of 6-mercaptopurine supplied as the hydrochloride salt for use as a tracer and internal standard in quantitative analytical methods. It is intended for NMR, GC-MS, and LC-MS workflows where isotopic labeling improves measurement accuracy.
  • Stable isotope-labeled 6-mercaptopurine hydrochloride, 13C and 15N labeled.
  • Suitable as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • High purity (≈99.9%), appropriate for quantitative analysis.
  • Supplied in small mg quantities for analytical use; store under nitrogen, away from moisture.
Catalog No. 50-002-84822