Learn More
Medchemexpress LLC 6-Methoxy-2-naphthoic acid | 2471-70-7 | 99.7% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB2121100G
Encompass_Preferred
6-Methoxy-2-naphthoic acid (6-MNA) is the active metabolite of Nabumetone. It acts as an inhibitor of COX-1 and COX-2, reducing the synthesis of gastric mucosal prostaglandin E2. This compound is valuable in the research of inflammation and pain-related diseases, and it also serves as an impurity in Naproxen.
  • Active metabolite.
  • Inhibits COX-1 and COX-2.
  • Inhibits synthesis of gastric mucosal prostaglandin E2.
  • Used in research of inflammation and pain-related diseases.
  • Serves as an impurity in some preparations.
Catalog No. 50-004-11488