Learn More
Cayman Chemical 6-methoxy-2-NaphthoIc AcId 25g
SDP
Supplier:  Cayman Chemical 3537425 G
Encompass_Preferred
An active metabolite of nabumetone; inhibits COX-1 and LPS-induced COX-2 activities in isolated human whole blood (IC50s = 31.01 and 19.84 µM, respectively); a potential impurity in commercial preparations of naproxen
Catalog No. 50-291-3298