Learn More
Medchemexpress LLC 6-methyl-1,4-naphthoquinone | 605-93-6 | MFCD04039611 | 98.5% | 172.18 | C11H8O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW58594350MG
Encompass_Preferred
6-Methyl-1,4-naphthoquinone is a small-molecule biochemical reagent used in organic synthesis and laboratory research. It is a solid compound with formula C11H8O2 and molecular weight 172.18 g/mol, commonly applied in redox chemistry, enzyme assays, and as a synthetic intermediate.
  • High purity: 98.5% (HPLC) with certificate of analysis available.
  • Identified by CAS number 605-93-6 and well characterized analytically.
  • Available in small-scale laboratory quantities for research use.
  • Supplied with SDS and COA for safety and quality verification.
  • Suitable for redox studies, enzymatic assays, and synthetic chemistry applications.
Catalog No. 50-002-80472