Learn More
Medchemexpress LLC 6-Methylsalicylic acid | 567-61-3 | MFCD00002166 | 152.15 | 50 G

Supplier: Medchemexpress LLC HYW00275150G
6-Methylsalicylic acid is a secondary metabolite isolated from certain fungi, such as Penicillium patulum. This compound is widely utilized as a building block for the synthesis of antibacterial and anti-inflammatory agents in various research applications.
- Isolated from fungi
- Building block for antibacterial agents
- Building block for anti-inflammatory agents
- Appearance: Light yellow to brown solid
- Melting point: 170-171°C
- Soluble in DMSO (100 mg/mL)
- Molecular formula: C8H8O3
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.