Learn More
Medchemexpress LLC 6-methyluracil | 626-48-2 | MFCD00006028 | 99.7% | 126.11 | C5H6N2O2 | 50g
SDP
Supplier:  Medchemexpress LLC HYY112550G
Encompass_Preferred
6-Methyluracil (Pseudothymine) a metabolite of Uracil can be used as an indicator of acetoacetyl-CoA (AACoA) accumulation 6-Methyluracil exhibits antiradiation effect in vivo[1][2]
Catalog No. 50-002-67097