Learn More
Medchemexpress LLC 6-Methyluridine | 16710-13-7 | C10H14N2O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1527325MG
Encompass_Preferred
6-Methyluridine is a uridine analogue with potential antiepileptic effects. Its analogs are used in research to study anticonvulsant, anxiolytic, and antihypertensive activities. This product is for research use only and shows cytotoxicity against various cell lines.
  • Uridine analogue
  • Potential antiepileptic effects
  • Studies anticonvulsant and anxiolytic activities
  • Develops new antihypertensive agents
  • Shows cytotoxicity against various cell lines
  • White to off-white solid appearance
Catalog No. 50-003-72271