Learn More
Medchemexpress LLC 6-(N-carbamoyl-2,6-difluoroanilino)-2-(2,4-difluorophenyl)pyridine-3-carboxamide | 745833-23-2 | MFCD11616590 | 99.7% | 404.32 g/mol | C19H12F4N4O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1040110MM 1ML
Encompass_Preferred
VX-702 is a selective small-molecule inhibitor of p38α mitogen-activated protein kinase (MAPK) used in research to probe p38α-mediated signaling and inflammatory pathways. It demonstrates higher potency for p38α than p38β and has been evaluated in preclinical and clinical studies for anti-inflammatory applications. Supplied for laboratory research use only.
  • Selective p38α MAPK inhibitor with approximately 14-fold selectivity over p38β.
  • Suitable for studying inflammatory signaling and cytokine regulation.
  • Provided as a 10 mM solution in DMSO for ready-to-use applications.
  • High purity (product page lists 99.69%), appropriate for biochemical assays.
  • Molecular weight 404.32 g/mol and formula C19H12F4N4O2.
  • Good solubility in DMSO (≥ 42 mg/mL) for convenient stock preparation.
  • For research use only; not intended for human or veterinary use.
Catalog No. 50-002-42486