Learn More
Medchemexpress LLC 6β-naltrexol-d3 | 1435727-11-9 | 346.44 | C20H22D3NO4 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY108229S500UG
Encompass_Preferred
6β-Naltrexol-d3 (6β-hydroxynaltrexone-d3) is a deuterium-labeled stable isotope of 6β-naltrexol used as an analytical reference standard and as an internal standard for quantitative mass spectrometry and tracer studies. It has molecular formula C20H22D3NO4 and molecular weight 346.44.
Catalog No. 50-002-41776