Learn More
Medchemexpress LLC 6"-O-Acetyldaidzin | 71385-83-6 | 5 MG

Supplier: Medchemexpress LLC HYN40715MG
6"-O-Acetyldaidzin is an isoflavone glycoside isolated from soybeans. It significantly inhibits lipid peroxidation in rat liver microsome with an IC50 of 8.2 μM.
- Purity of 99.89%
- Molecular weight of 458.41
- Chemical formula C23H22O10
- Appearance as a solid
- Color is off-white to light yellow
- Soluble in DMSO at 4.6 mg/mL (10.03 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.