Learn More
Medchemexpress LLC 6-O-Methyl-guanine | 20535-83-5 | 98.9% | 165.16 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW013591
Encompass_Preferred
6-O-Methyl-guanine is a biochemical reagent suitable for life science related research. It is intended for research use only.
  • Solid, white to off-white appearance
  • Ships at room temperature in the continental US
  • Store at 4°C, protected from light
  • For storage in solvent, keep at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-51469