Learn More
Medchemexpress LLC 6-oxolithocholic acid | 2393-61-5 | MFCD00070044 | 100.0% | 390.56 | C24H38O4 | 1 MG

Supplier: Medchemexpress LLC HYW5875301MG
6-Oxolithocholic acid is a bile acid metabolite provided as a research-grade solid for biochemical and cell biology studies. It is commonly used to investigate bile acid metabolism, cytotoxicity, and apoptosis pathways in vitro, and is supplied with documented purity and storage recommendations.
- High purity: 99.99% (manufacturer stated)
- Form: white to off-white solid powder
- Molecular formula: C24H38O4; molecular weight: 390.56
- Applications: studies of bile acid metabolism, cytotoxicity, and apoptosis in vitro
- Storage: powder stable at -20 °C for long-term storage
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.