Learn More
Medchemexpress LLC 6-phosphoglucono-δ-lactone | 2641-81-8 | 99.0% | 258.12 g/mol | C6H11O9P | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW5877961MG
Encompass_Preferred
6-Phosphoglucono-δ-lactone is a biochemical intermediate in the oxidative branch of the pentose phosphate pathway and functions as a reversal substrate of glucose-6-phosphate dehydrogenase (G6PD). It is produced from D-glucose-6-phosphate by G6PDH and is converted to 6-phosphogluconate by lactonase. Supplied for research applications including studies of metabolism, heart failure, type 2 diabetes, and cancer.
  • High purity (99.0%) for research use.
  • Small-quantity packaging suitable for assay development and enzymatic studies.
  • Biochemically relevant substrate for studying G6PD activity and the pentose phosphate pathway.
  • Water-soluble phosphorylated lactone useful as a metabolic intermediate probe.
Catalog No. 50-002-85032