Learn More
USA Scientific Inc 6-place (1 x 6) chest freezer rack for 4 3/4 inch H boxes
Supplier:  USA Scientific Inc 26902306
Encompass
6-place (1 long x 6 high) chest freezer rack for 4 3/4 in H boxes, 6 in L x 6 1/8 in W x 29 5/8 in H, stainless steel
Catalog No. 50-754-1185