Learn More
Electron Microscopy Sciences 6" Polypropylene Shaft Foam Over Cotton
SDP
Supplier:  Electron Microscopy Sciences 72326
Encompass_Preferred
Tips withstand many chemicals and solutions used in labs and clean rooms. Resistant to deterioration. Head is made from 100% urethane foam. 6" polypropylene shaft, foam-over-cotton, tip: 3⁄8"Dia x 15⁄16"
Catalog No. 50-949-193